missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dibromopentane, 97%, Thermo Scientific Chemicals

Product Code. 11302297
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1,4-Dibromopentane is used in the preparation of pharaoh ant trail pheromone stereoisomer, indolizidine alkaloid. It is also involved in the preparation 1,2-dimethyl-1-phenylcyclopentane and boc-L-2-amino-6-bromoheptanoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dibromopentane is used in the preparation of pharaoh ant trail pheromone stereoisomer, indolizidine alkaloid. It is also involved in the preparation 1,2-dimethyl-1-phenylcyclopentane and boc-L-2-amino-6-bromoheptanoic acid.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-87-9
Molecular Formula C5H10Br2
Molecular Weight (g/mol) 229.943
MDL Number MFCD00000153
InChI Key CNBFRBXEGGRSPL-UHFFFAOYSA-N
Synonym pentane, 1,4-dibromo, 1,4-dibrompentan, pentane,4-dibromo, 1,4-dibromo-pentane, pubchem12546, 1,4-dibromopentane, hexanoic acid monoethyl ester
PubChem CID 79082
IUPAC Name 1,4-dibromopentane
SMILES CC(CCCBr)Br

Specifications

Melting Point -34°C
Density 1.687
Boiling Point 102°C to 103°C (15 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5085
Quantity 100 g
Beilstein 605295
Solubility Information Insoluble in water.
Formula Weight 229.95
Percent Purity 97%
Chemical Name or Material 1,4-Dibromopentane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'