missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dichlorobutane, 99%, Thermo Scientific Chemicals

Product Code. 10471831
Click to view available options
Quantity:
5 mL
100 mL
500 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-56-5
Molecular Formula C4H8Cl2
Molecular Weight (g/mol) 127.01
MDL Number MFCD00001011
InChI Key KJDRSWPQXHESDQ-UHFFFAOYSA-N
Synonym butane, 1,4-dichloro, unii-8326ynm2b3, tetramethylene chloride, 1,4-dichlorobutane, butane,4-dichloro, tetramethylene dichloride, 1,4-bis chloranyl butane, ksc491k8b, 1,4-dichlorobutane, puriss gc
PubChem CID 8059
IUPAC Name 1,4-dichlorobutane
SMILES ClCCCCCl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -38°C
Density 1.1600g/mL
Boiling Point 161°C to 163°C
Flash Point 40°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula Cl(CH2)4Cl
Refractive Index 1.4530 to 1.455
Quantity 100 mL
Beilstein 01, 119
Specific Gravity 1.16
Solubility Information Solubility in water: 0.24% (20 c). Other solubilities: soluble in most common organic solvents
Viscosity 1.46 mPa.s (20°C)
Formula Weight 127.01
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1, 4-Dichlorobutane, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • 1,4-Dichlorobutane
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Flammable liquid Category 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'