missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dimethoxynaphthalene, 96%, Thermo Scientific Chemicals

Product Code. 11408046
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1,4-Dimethoxynaphthalene may be used as a reference molecule during the synthesis, separation and analysis of derivatized naphthalenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dimethoxynaphthalene may be used as a reference molecule during the synthesis, separation and analysis of derivatized naphthalenes.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10075-62-4
Molecular Formula C12H12O2
Molecular Weight (g/mol) 188.226
MDL Number MFCD00052378
InChI Key FWWRTYBQQDXLDD-UHFFFAOYSA-N
PubChem CID 82337
IUPAC Name 1,4-dimethoxynaphthalene
SMILES COC1=CC=C(C2=CC=CC=C21)OC

Specifications

Melting Point 84°C to 85°C
Quantity 1 g
Beilstein 2048082
Synonym naphthalene, 1,4-dimethoxy, 1,4-dimethoxy-naphthalene, maybridge1_003006, naphthalene,4-dimethoxy, acmc-1c6fs, 1,4-dimethoxy naphthalene, naphthalene,1,4-dimethoxy, 1,4-dimethoxynaphthalene, 1,4-dimethoxynaphthalene, for fluorescence, fluorescent marker hplc
Solubility Information Soluble in water.
Formula Weight 188.23
Percent Purity 96%
Chemical Name or Material 1,4-Dimethoxynaphthalene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'