missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dioxane, 99+%, extra pure, stabilized, Thermo Scientific Chemicals

100.50 € - 1188.92 €

Chemical Identifiers

CAS 123-91-1
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00006571
InChI Key RYHBNJHYFVUHQT-UHFFFAOYSA-N
Synonym dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
PubChem CID 31275
ChEBI CHEBI:47032
IUPAC Name 1,4-dioxane
SMILES C1COCCO1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10294520
View Documents
Thermo Scientific Acros
117110010
1 L Glass Bottle
100.50 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10604772
View Documents
Thermo Scientific Acros
117110025
2.5 L Glass Bottle
188.94 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10468450
View Documents
Thermo Scientific Acros
117110250
25 L Metal Drum
1188.92 €
25 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

123-91-1
88.106
RYHBNJHYFVUHQT-UHFFFAOYSA-N
31275
1,4-dioxane
C4H8O2
MFCD00006571
dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
CHEBI:47032
C1COCCO1

Specifications

12.0°C
99.0
100.0
101.0°C
Liquid
99+%
1.034
MFCD00006571
19, 3
01,333
dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
RYHBNJHYFVUHQT-UHFFFAOYSA-N
1,4-dioxane
31275
88.11
Extra Pure
123-91-1
Colorless
1.0340g/mL
12°C
1.4205 to 1.4220
0.01% max.
C4H8O2
1.32 mPa.s (20°C)
0.1% max.
15, 3328
Solubility in water: soluble. Other solubilities: soluble in ethanol, acetone, benzene, toluene,, ether and other common organic solvents
C1COCCO1
88.106
CHEBI:47032
≥99%
1, 4-Dioxane, 99+%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Suspected of causing cancer.
May cause respiratory irritation.
Highly flammable liquid and vapor.
May form explosive peroxides.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep container tightly closed.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

missing translation for 'einecsNumber' : 204-661-8

missing translation for 'rtecsNumber' : JG8225000

missing translation for 'tsca' : TSCA

Product Identifier
  • 1,4-Dioxane

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Carcinogenicity Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only