missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dioxane, anhydrous, 99.8%, stab. with 1-3ppm BHT, packaged under argon in resealable AcroSeal™ bottles

Product Code. p-7232518
Click to view available options
Quantity:
100 mL
1 L
4 x 1 L
Unit Size:
1 L
100mL
4 L
This item is not returnable. View return policy

Product Code. 16368207

Brand: Thermo Scientific Alfa Aesar 047305.AE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123-91-1 | C{4}H{8}O{2}

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 11.8°C
CAS 123-91-1
Boiling Point 100°C to 102°C
Quantity 100 mL
Molecular Formula C{4}H{8}O{2}
Percent Purity 99.8%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.