missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Dioxane, Certified AR for Analysis, Stabilised with BHT, Fisher Chemical™
66.40 € - 1030.00 €
Chemical Identifiers
CAS | 123-91-1 |
---|---|
Molecular Formula | C4H8O2 |
Molecular Weight (g/mol) | 88.106 |
MDL Number | 6571 |
InChI Key | RYHBNJHYFVUHQT-UHFFFAOYSA-N |
Synonym | dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin |
PubChem CID | 31275 |
ChEBI | CHEBI:47032 |
IUPAC Name | 1,4-dioxane |
SMILES | C1COCCO1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11413733
|
Fisher Chemical
D/4550/08 |
500 mL | Amber Glass Bottle |
66.40 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10141470
|
Fisher Chemical
D/4550/15 |
1 L | Amber Glass Bottle |
108.00 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10000712
|
Fisher Chemical
D/4550/PB15 |
1 L | Plastic-coated Glass Bottle |
117.00 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10080120
|
Fisher Chemical
D/4550/PB17 |
2.5 L | Plastic-coated Glass Bottle |
282.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10651711
|
Fisher Chemical
D/4550/25 |
25 L | Metal Can |
1030.00 €
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
1,4-Dioxane, Certified AR for Analysis, Stabilised with BHT
Chemical Identifiers
123-91-1 | |
88.106 | |
RYHBNJHYFVUHQT-UHFFFAOYSA-N | |
31275 | |
1,4-dioxane |
C4H8O2 | |
6571 | |
dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin | |
CHEBI:47032 | |
C1COCCO1 |
Specifications
6 to 8 | |
123-91-1 | |
101°C | |
C4H8O2 | |
41mbar at 20°C | |
dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin | |
C1COCCO1 | |
88.106 | |
CHEBI:47032 |
12°C | |
Colorless | |
Liquid | |
6571 | |
1.32 mPaS at 20°C | |
RYHBNJHYFVUHQT-UHFFFAOYSA-N | |
1,4-dioxane | |
31275 | |
88.11g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 1,4-Dioxane
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Carcinogenicity Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- EUH019-May form explosive peroxides.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P233-Keep container tightly closed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.