missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dioxane, Extra Pure, SLR, Stabilised with BHT, Fisher Chemical™

47.40 € - 615.00 €

Chemical Identifiers

CAS 123-91-1
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number 6571
InChI Key RYHBNJHYFVUHQT-UHFFFAOYSA-N
Synonym dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
PubChem CID 31275
ChEBI CHEBI:47032
IUPAC Name 1,4-dioxane
SMILES C1COCCO1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10060120
View Documents Promotion Details
Fisher Chemical
D/4500/08
500 mL Amber Glass Bottle
47.40 €
500mL
Please to purchase this item. Need a web account? Register with us today!
10112190
View Documents Promotion Details
Fisher Chemical
D/4500/21
5 L Metal Can
208.00 €
5 L
Please to purchase this item. Need a web account? Register with us today!
10070120
View Documents Promotion Details
Fisher Chemical
D/4500/25
25 L Metal Can
615.00 €
25 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

123-91-1
88.106
RYHBNJHYFVUHQT-UHFFFAOYSA-N
31275
1,4-dioxane
C4H8O2
6571
dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
CHEBI:47032
C1COCCO1

Specifications

6 to 8
123-91-1
101°C
C4H8O2
41mbar at 20°C
dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
C1COCCO1
88.106
CHEBI:47032
12°C
Colorless
Liquid
6571
1.32 mPaS at 20°C
RYHBNJHYFVUHQT-UHFFFAOYSA-N
1,4-dioxane
31275
88.11g/mol
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 1,4-Dioxane

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Carcinogenicity Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
  • EUH019-May form explosive peroxides.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P233-Keep container tightly closed.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : 1,4-Dioxane

SDS
Documents

Documents

Product Certifications