missing translation for 'onlineSavingsMsg'
Learn More

1,4-dioxaspiro[4.5]decan-8-one, Thermo Scientific™

Product Code. 10651455
Click to view available options
Quantity:
10 g
25 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 4746-97-8
Molecular Formula C8H12O3
Molecular Weight (g/mol) 156.181
InChI Key VKRKCBWIVLSRBJ-UHFFFAOYSA-N
Synonym 1,4-dioxaspiro 4.5 decan-8-one, 1,4-cyclohexanedione monoethylene acetal, 1,4-cyclohexanedione monoethylene ketal, 1,4-cyclohexanedione monoethyleneacetal, 1,4-dioxa-spiro 4.5 decan-8-one, 1,4-cyclohexanedione monoethyleneketal, cyclohexane-1,4-dione monoethylene ketal, 1,4-dioxaspiro 4.5 decane-8-one
PubChem CID 567415
IUPAC Name 1,4-dioxaspiro[4.5]decan-8-one
SMILES C1CC2(CCC1=O)OCCO2

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 156.18
Chemical Name or Material 1, 4-Dioxaspiro[4.5]decan-8-one
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H335-May cause respiratory irritation.
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Cyclohexane-1,4-dione, monoethylene ketal
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'