missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Ethoxyphenyl)ethanol, 98%, Thermo Scientific™

27.40 € - 289.00 €

Chemical Identifiers

CAS 52067-36-4
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD08062382
InChI Key GKGQWBJLOYXULB-UHFFFAOYSA-N
Synonym 1-4-ethoxyphenyl ethan-1-ol, 1-4-ethoxyphenyl ethanol
PubChem CID 13891147
IUPAC Name 1-(4-ethoxyphenyl)ethanol
SMILES CCOC1=CC=C(C=C1)C(C)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15403857
View Documents
Thermo Scientific Alfa Aesar
H30052.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15413857
View Documents
Thermo Scientific Alfa Aesar
H30052.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15423857
View Documents
Thermo Scientific Alfa Aesar
H30052.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

52067-36-4
166.22
GKGQWBJLOYXULB-UHFFFAOYSA-N
13891147
CCOC1=CC=C(C=C1)C(C)O
C10H14O2
MFCD08062382
1-4-ethoxyphenyl ethan-1-ol, 1-4-ethoxyphenyl ethanol
1-(4-ethoxyphenyl)ethanol

Specifications

52067-36-4
C10H14O2
1-4-ethoxyphenyl ethan-1-ol, 1-4-ethoxyphenyl ethanol
CCOC1=CC=C(C=C1)C(C)O
166.22
166.22
1-(4-Ethoxyphenyl)ethanol
46°C to 47°C
MFCD08062382
GKGQWBJLOYXULB-UHFFFAOYSA-N
1-(4-ethoxyphenyl)ethanol
13891147
98%
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.