missing translation for 'onlineSavingsMsg'
Learn More

[1-(4-Fluorophenyl)-5-methyl-1H-pyrazol-4-yl]methanol, 97%, Thermo Scientific™

Product Code. 10354242
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 465514-37-8
Molecular Formula C11H11FN2O
Molecular Weight (g/mol) 206.22
MDL Number MFCD03644148
InChI Key QJPGMVFNIWHOIY-UHFFFAOYSA-N
Synonym 1-4-fluorophenyl-5-methyl-1h-pyrazol-4-yl methanol, 1-4-fluorophenyl-5-methylpyrazol-4-yl methanol, fmq, maybridge3_004281, 1-4-fluorophenyl-5-methyl-4-pyrazolyl methanol, 1-4-fluorophenyl-5-methyl-pyrazol-4-yl methanol, 1-4-fluorophenyl-5-methyl-1h-pyrazole-4-methanol, 1-4-fluorophenyl-5-methylpyrazol-4-yl methan-1-ol
PubChem CID 2780717
IUPAC Name [1-(4-fluorophenyl)-5-methylpyrazol-4-yl]methanol
SMILES CC1=C(CO)C=NN1C1=CC=C(F)C=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 206.22
Percent Purity 97%
Chemical Name or Material [1-(4-Fluorophenyl)-5-methyl-1H-pyrazol-4-yl]methanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.