missing translation for 'onlineSavingsMsg'
Learn More

[1-(4-Fluorophenyl)-5-methyl-1H-pyrazol-4-yl]methanol, 97%, Thermo Scientific™

Product Code. 10752873
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 465514-37-8
Molecular Formula C11H11FN2O
Molecular Weight (g/mol) 206.22
MDL Number MFCD03644148
InChI Key QJPGMVFNIWHOIY-UHFFFAOYSA-N
Synonym 1-4-fluorophenyl-5-methyl-1h-pyrazol-4-yl methanol, 1-4-fluorophenyl-5-methylpyrazol-4-yl methanol, fmq, maybridge3_004281, 1-4-fluorophenyl-5-methyl-4-pyrazolyl methanol, 1-4-fluorophenyl-5-methyl-pyrazol-4-yl methanol, 1-4-fluorophenyl-5-methyl-1h-pyrazole-4-methanol, 1-4-fluorophenyl-5-methylpyrazol-4-yl methan-1-ol
PubChem CID 2780717
IUPAC Name [1-(4-fluorophenyl)-5-methylpyrazol-4-yl]methanol
SMILES CC1=C(CO)C=NN1C1=CC=C(F)C=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 206.22
Percent Purity 97%
Chemical Name or Material [1-(4-Fluorophenyl)-5-methyl-1H-pyrazol-4-yl]methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'