missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Nitrophenyl)piperazine, 98%, Thermo Scientific Chemicals

Product Code. 11329475
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6269-89-2
Molecular Formula C10H13N3O2
Molecular Weight (g/mol) 207.233
MDL Number MFCD00005961
InChI Key VWOJSRICSKDKAW-UHFFFAOYSA-N
Synonym 1-4-nitrophenyl piperazine, 1-4-nitro-phenyl-piperazine, piperazine, 1-4-nitrophenyl, 1-4-nitrophenyl-piperazine, n-4-nitrophenyl piperazine, n-4-nitrophenyl-piperazine, 4-nitrophenyl piperazine, 1-4-nitrophenyl piperazin, pubchem8580, 4-nitrophenylpiperazine
PubChem CID 80447
SMILES C1CN(CCN1)C2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Melting Point 129°C to 133°C
Quantity 5 g
UN Number UN2811
Formula Weight 207.23
Percent Purity ≥98%
Chemical Name or Material 1-(4-Nitrophenyl)piperazine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'