missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Piperidyl)-1H-1,2,3-benzotriazole hydrochloride, ≥90%, Thermo Scientific™

Product Code. 10151122
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 79098-80-9
Molecular Formula C11H15ClN4
Molecular Weight (g/mol) 238.719
MDL Number MFCD00831110
InChI Key SIZIQJKLJZWLLD-UHFFFAOYSA-N
Synonym 1-4-piperidyl-1h-1,2,3-benzotriazole hydrochloride, 1-piperidin-4-ylbenzotriazole hydrochloride, 1-piperidin-4-yl benzotriazole hydrochloride, 1-piperidin-4-yl-1h-benzotriazole hydrochloride, 1-piperidin-4-yl-1,2,3-benzotriazole hydrochloride, c11h14n4.hcl, 4-piperidylbenzotriazole, chloride, 4-benzotriazol-1-yl-piperidine hcl salt, 4-benzotriazol-1-ylpiperidine hydrochloride, 1-4-piperidinyl benzotriazole hydrochloride
PubChem CID 2779380
IUPAC Name 1-piperidin-4-ylbenzotriazole;hydrochloride
SMILES C1CNCCC1N2C3=CC=CC=C3N=N2.Cl

Specifications

CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 238.72
Percent Purity 90%
Chemical Name or Material 1-(4-Piperidyl)-1H-1, 2, 3-benzotriazole hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'