missing translation for 'onlineSavingsMsg'
Learn More

1-(5-Chloro-2-hydroxyphenyl)-3-phenyl-1,3-propanedione, 98%, Thermo Scientific™

Product Code. 10535643
Click to view available options
1g; Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 5067-25-4
CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C15H11ClO3
Linear Formula C6H5COCH2COC6H3(Cl)OH
MDL Number MFCD00209562
Quantity 1g
Synonym 1-5-chloro-2-hydroxyphenyl-3-phenylpropane-1,3-dione, 1-5-chloro-2-hydroxyphenyl-3-phenyl-1,3-propanedione, 1-5-chloranyl-2-oxidanyl-phenyl-3-phenyl-propane-1,3-dione, cambridge id 5479511, regid_for_cid_724088, 1,3-propanedione,1-5-chloro-2-hydroxyphenyl-3-phenyl, 1-2-hydroxy-5-chlorophenyl-3-phenyl-1,3-propanedione, 1-5-chloro-2-hydroxy-phenyl-3-phenyl-propane-1,3-dione
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform,ethanol,ethylacetate
InChI Key VOVLKGWYQQCQHB-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C(=O)CC(=O)C2=C(C=CC(=C2)Cl)O
IUPAC Name 1-(5-chloro-2-hydroxyphenyl)-3-phenylpropane-1,3-dione
Molecular Weight (g/mol) 274.7
PubChem CID 724088
Formula Weight 274.7
Percent Purity 98%
Chemical Name or Material 1-(5-Chloro-2-hydroxyphenyl)-3-phenyl-1, 3-propanedione
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'