missing translation for 'onlineSavingsMsg'
Learn More

1,5-Diamino-4,8-dihydroxy-9,10-dihydroanthracene-9,10-dione, Tech., Thermo Scientific™

Product Code. 10554695
Click to view available options
Quantity:
10 g
50 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 145-49-3
Molecular Formula C14H10N2O4
Molecular Weight (g/mol) 270.244
MDL Number MFCD00013722
InChI Key HSYLKWSCFRLSKB-UHFFFAOYSA-N
Synonym 1,5-diamino-4,8-dihydroxyanthraquinone, diaminoanthrarufin, 1,5-diaminoanthrarufin, 4,8-diaminoanthrarufin, 1,5-dihydroxy-4,8-diaminoanthraquinone, anthrarufin, 4,8-diamino, unii-q7yvg56q18, ccris 5599, 4,8-diamino-1,5-dihydroxyanthraquinone, 1,5-diamino-4,8-dihydroxy-anthracene-9,10-dione
PubChem CID 67351
IUPAC Name 1,5-diamino-4,8-dihydroxyanthracene-9,10-dione
SMILES C1=CC(=C2C(=C1N)C(=O)C3=C(C=CC(=C3C2=O)N)O)O

Specifications

Quantity 100 g
Formula Weight 270.24
Percent Purity Tech.
Grade Technical
Chemical Name or Material 1,5-Diamino-4,8-dihydroxy-9,10-dihydroanthracene-9,10-dione
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.