missing translation for 'onlineSavingsMsg'
Learn More

1,5-Diaminopentane, 98%, Thermo Scientific Chemicals

Product Code. 11466226
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1,5-Diaminopentane is mainly used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,5-Diaminopentane is mainly used in organic synthesis.

Solubility
Soluble in water.

Notes
Air sensitive & hygroscopic. Incompatible with acid chlorides, acids, acid anhydrides, strong oxidizing agents, carbon dioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 462-94-2
Molecular Formula C5H14N2
Molecular Weight (g/mol) 102.18
MDL Number MFCD00008239
InChI Key VHRGRCVQAFMJIZ-UHFFFAOYSA-N
Synonym 1,5-diaminopentane, cadaverine, 1,5-pentanediamine, pentamethylenediamine, cadaverin, animal coniine, 1,5-pentamethylenediamine, unii-l90ben6oll, l90ben6oll, n2p
PubChem CID 273
ChEBI CHEBI:18127
IUPAC Name pentane-1,5-diamine
SMILES NCCCCCN

Specifications

Melting Point 9°C
Density 0.873
Boiling Point 178°C to 180°C
Assay Percent Range 0.98
Odor Amine-like
Linear Formula H2N(CH2)5NH2
Refractive Index 1.4585
Quantity 25 g
UN Number UN2735
Sensitivity Air sensitive; Hygroscopic
Merck Index 14,1609
Solubility Information Soluble in water.
Formula Weight 102.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1,5-Diaminopentane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.