missing translation for 'onlineSavingsMsg'
Learn More

1,5-Diazabicyclo[4.3.0]non-5-ene, 98%

Product Code. 11475483
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11475483

Brand: Thermo Scientific Alfa Aesar A13437.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3001-72-7 | C7H12N2 | 124.19 g/mol

1,5-Diazabicyclo[4.3.0]non-5-ene is employed for dehydrohalogenation reactions and base-catalyzed rearrangements in organic synthesis. It is an amidine base used in organic synthesis. Further, it is used as a resin curing agent and polyurethane catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,5-Diazabicyclo[4.3.0]non-5-ene is employed for dehydrohalogenation reactions and base-catalyzed rearrangements in organic synthesis. It is an amidine base used in organic synthesis. Further, it is used as a resin curing agent and polyurethane catalyst.

Solubility
Miscible with water.

Notes
Air, light and moisture sensitive. Hygroscopic. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acid chlorides, acid anhydrides, oxidizing agents and chloroformates.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3001-72-7
Molecular Formula C7H12N2
Molecular Weight (g/mol) 124.19
MDL Number MFCD00005554
InChI Key SGUVLZREKBPKCE-UHFFFAOYSA-N
Synonym 1,5-diazabicyclo 4.3.0 non-5-ene, 2,3,4,6,7,8-hexahydropyrrolo 1,2-a pyrimidine, unii-978m4ol12q, 2h,3h,4h,6h,7h,8h-pyrrolo 1,2-a pyrimidine, pyrrolo 1,2-a pyrimidine, 2,3,4,6,7,8-hexahydro, 1,5-diazabicyclo 4,3,0 non-5-ene, 1,5-diazabicyclo 4.3.0-5-nonene, 1,5-diazabicyclo-4.3.0 non-5-ene
PubChem CID 76349
IUPAC Name 2,3,4,6,7,8-hexahydropyrrolo[1,2-a]pyrimidine
SMILES C1CN2CCCN=C2C1

Specifications

Density 1.042
Boiling Point 95°C to 98°C (7 mmHg)
Flash Point 94°C (201°F)
Refractive Index 1.52
Quantity 25 g
UN Number UN3267
Beilstein 2417
Sensitivity Air sensitive; Hygroscopic
Solubility Information Miscible with water.
Formula Weight 124.19
Percent Purity 98%
Chemical Name or Material 1,5-Diazabicyclo[4.3.0]non-5-ene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.