missing translation for 'onlineSavingsMsg'
Learn More
1,5-Diazabicyclo[4.3.0]non-5-ene, 98%, Thermo Scientific Chemicals Product Code.: 10265630

1,5-Diazabicyclo[4.3.0]non-5-ene, 98%, Thermo Scientific Chemicals

Product Code. 10265630
100 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10265630

Brand: Thermo Scientific Acros 159611000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3001-72-7
Molecular Formula C7H12N2
Molecular Weight (g/mol) 124.19
MDL Number MFCD00005554
InChI Key SGUVLZREKBPKCE-UHFFFAOYSA-N
Synonym 1,5-diazabicyclo 4.3.0 non-5-ene, 2,3,4,6,7,8-hexahydropyrrolo 1,2-a pyrimidine, unii-978m4ol12q, 2h,3h,4h,6h,7h,8h-pyrrolo 1,2-a pyrimidine, pyrrolo 1,2-a pyrimidine, 2,3,4,6,7,8-hexahydro, 1,5-diazabicyclo 4,3,0 non-5-ene, 1,5-diazabicyclo 4.3.0-5-nonene, 1,5-diazabicyclo-4.3.0 non-5-eneShow MoreShow Less
PubChem CID 76349
IUPAC Name 2,3,4,6,7,8-hexahydropyrrolo[1,2-a]pyrimidine
SMILES C1CN2CCCN=C2C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0000g/mL
Boiling Point 95.0°C to 98.0°C (7.5 mmHg)
Flash Point 94°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5190 to 1.5210
Quantity 100 g
Fieser 01,189; 02,98; 04,116; 05,176; 06,157; 07,86; 13,91
Merck Index 15, 2995
Specific Gravity 1
Solubility Information Solubility in water: soluble
Formula Weight 124.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1,5-Diazabicyclo[4.3.0]non-5-ene, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0427437 Certificate TypeCertificate of Analysis Date12/03/2024 Catalog Number159610250,159611000
Lot NumberA0449089 Certificate TypeCertificate of Analysis Date06/01/2023 Catalog Number159610250,159611000
Lot NumberA0399570 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number159610250,159611000
Lot NumberA0410552 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number159610250,159611000
Lot NumberA0447286 Certificate TypeCertificate of Analysis Date06/12/2022 Catalog Number159610250,159611000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,5-Diazabicyclo[4.3.0]non-5-ene, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.