missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dibromo-3-methylpentane, 98+%, Thermo Scientific Chemicals

76.58 € - 306.53 €

Chemical Identifiers

CAS 4457-72-1
Molecular Formula C6H12Br2
Molecular Weight (g/mol) 243.97
MDL Number MFCD00039200
InChI Key YDPZWUMQKMLLHC-UHFFFAOYSA-N
Synonym 3-methyl-1,5-dibromopentane, 1,5-dibromo-3-methyl-pentane, pentane, 1,5-dibromo-3-methyl, acmc-20ap1m, pentane,1,5-dibromo-3-methyl, 1,5-dibromo-3-methylpentane 5g
PubChem CID 138258
IUPAC Name 1,5-dibromo-3-methylpentane
SMILES CC(CCBr)CCBr
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11418887
View Documents
Thermo Scientific Alfa Aesar
B20521.06
5 g
76.58 €
5g
Please to purchase this item. Need a web account? Register with us today!
11428887
View Documents
Thermo Scientific Alfa Aesar
B20521.14
25 g
306.53 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

1,5-Dibromo-3-methylpentane is used as a chemical reagent, pharmaceutical intermediate, pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,5-Dibromo-3-methylpentane is used as a chemical reagent, pharmaceutical intermediate, pharmaceutical research.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, strong bases.
Specifications

Chemical Identifiers

4457-72-1
243.97
YDPZWUMQKMLLHC-UHFFFAOYSA-N
138258
CC(CCBr)CCBr
C6H12Br2
MFCD00039200
3-methyl-1,5-dibromopentane, 1,5-dibromo-3-methyl-pentane, pentane, 1,5-dibromo-3-methyl, acmc-20ap1m, pentane,1,5-dibromo-3-methyl, 1,5-dibromo-3-methylpentane 5g
1,5-dibromo-3-methylpentane

Specifications

4457-72-1
233°C
C6H12Br2
1.508
3-methyl-1,5-dibromopentane, 1,5-dibromo-3-methyl-pentane, pentane, 1,5-dibromo-3-methyl, acmc-20ap1m, pentane,1,5-dibromo-3-methyl, 1,5-dibromo-3-methylpentane 5g
YDPZWUMQKMLLHC-UHFFFAOYSA-N
1,5-dibromo-3-methylpentane
138258
≥98%
1.58
101°C (214°F)
BrCH2CH2CH(CH3)CH2CH2Br
MFCD00039200
Insoluble in water.
CC(CCBr)CCBr
243.97
243.98
1,5-Dibromo-3-methylpentane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.