missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dimethyl-1H-pyrazole-3-carboxylic acid, 95+%, Thermo Scientific™

Product Code. 10442064
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
This item is not returnable. View return policy

Product Code. 10442064

Brand: Thermo Scientific Maybridge CC12601EA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 5744-59-2
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.14
MDL Number MFCD00085040
InChI Key PXRXGHUTKHXUGF-UHFFFAOYSA-N
Synonym 1,5-dimethyl-1h-pyrazole-3-carboxylic acid, 1,5-dimethyl-1h-pyrazole-3-carboxylicacid, 1,5-dimethyl-3-pyrazolecarboxylic acid, 1h-pyrazole-3-carboxylic acid, 1,5-dimethyl, 1,5-dimethyl-1h-pyrazole-3-carboxylic, acmc-209lyb, pyrazole-3-carboxylic acid, 1,5-dimethyl, 1h-pyrazole-3-carboxylicacid, 1,5-dimethyl, 1,5-dimethyl-1h-pyrazole-3-carboxylic acid #
PubChem CID 587757
SMILES CN1N=C(C=C1C)C(O)=O

Specifications

Assay Percent Range 95%
Quantity 10 g
IUPAC Name 1,5-dimethylpyrazole-3-carboxylic acid
Formula Weight 140.14
Percent Purity 95%
Chemical Name or Material 1, 5-Dimethyl-1H-pyrazole-3-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.