missing translation for 'onlineSavingsMsg'
Learn More

1,5-Diphenylcarbazide, ACS, Thermo Scientific Chemicals

Product Code. 11377458
Click to view available options
Quantity:
5 g
25 g
100 g

1,5-Diphenylcarbazide is used in the colorimetric determination of chromium, osmium as well as in the detection of cadmium, mercury, magnesium, emetine and aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140-22-7
Molecular Formula C13H14N4O
Molecular Weight (g/mol) 242.282
MDL Number MFCD00003013
InChI Key KSPIHGBHKVISFI-UHFFFAOYSA-N
Synonym 1,5-diphenylcarbazide, diphenylcarbazide, 1,5-diphenylcarbohydrazide, carbonic dihydrazide, 2,2'-diphenyl, 1,5-diphenylcarbonohydrazide, 1,5-diphenylcabohydrazide, sym-diphenylcarbazide, n,n'-diphenylcarbazide, 2,2'-diphenylcarbazide, carbohydrazide, 1,5-diphenyl
PubChem CID 8789
ChEBI CHEBI:4641
IUPAC Name 1,3-dianilinourea
SMILES C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2

Specifications

Melting Point 170°C to 176°C
Color White to Cream
Quantity 25 g
Beilstein 752039
Sensitivity Air and light sensitive
Merck Index 14,3321
Solubility Information Soluble in glacial acetic acid,alcohol and acetone; Slightly soluble in water.
Formula Weight 242.28
Grade ACS
Physical Form Crystalline
Chemical Name or Material 1,5-Diphenylcarbazide
Show More Show Less
compliance-icons
Product Identifier
  • 1,5-Diphenylcarbazide
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.