missing translation for 'onlineSavingsMsg'
Learn More

1-(5-Fluoro-2-methoxyphenyl)ethanol, 95%, Thermo Scientific™

Product Code. 15414697
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 878572-08-8
Molecular Formula C9H11FO2
Molecular Weight (g/mol) 170.18
MDL Number MFCD08062369
InChI Key VLBOHPCLXJIASU-UHFFFAOYNA-N
Synonym 1-5-fluoro-2-methoxyphenyl ethanol, 1-5-fluoro-2-methoxyphenyl ethan-1-ol, benzenemethanol,5-fluoro-2-methoxy-a-methyl, 1-5-fluoro-2-methoxy-phenyl-ethanol, 1-5-fluoro-2-methoxyphenyl ethan-1-ol_x000d_
PubChem CID 44891074
IUPAC Name 1-(5-fluoro-2-methoxyphenyl)ethanol
SMILES COC1=C(C=C(F)C=C1)C(C)O

Specifications

Quantity 5 g
Formula Weight 170.18
Percent Purity 95%
Chemical Name or Material 1-(5-Fluoro-2-methoxyphenyl)ethanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'