missing translation for 'onlineSavingsMsg'
Learn More
1,6-Anhydro-beta-D-glucopyranose, 99%, Thermo Scientific Chemicals Product Code.: 11492728

1,6-Anhydro-beta-D-glucopyranose, 99%, Thermo Scientific Chemicals

Product Code. 11492728
1 g
Click to view available options
Quantity:
0.25 g
1 g
This item is not returnable. View return policy

Product Code. 11492728

Brand: Thermo Scientific Alfa Aesar A16724.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1,6-Anhydro-beta-D-glucopyranose is used for the preparation of biologically important and structurally diverse products such as rifamycin S, indanomycin, thromboxane B2, (+)-biotin, tetrodotoxin, quinone, macrolide antibiotics and modified sugars. It is used as a chemical tracer for biomass burning in atmospheric chemistry studies, particularly with respect to airborne particulate matter. It aids in the production of bioethanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,6-Anhydro-beta-D-glucopyranose is used for the preparation of biologically important and structurally diverse products such as rifamycin S, indanomycin, thromboxane B2, (+)-biotin, tetrodotoxin, quinone, macrolide antibiotics and modified sugars. It is used as a chemical tracer for biomass burning in atmospheric chemistry studies, particularly with respect to airborne particulate matter. It aids in the production of bioethanol.

Solubility
Soluble in Ethanol and Water.

Notes
Store at -20°C. Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 498-07-7
Molecular Formula C6H10O5
Molecular Weight (g/mol) 162.14
MDL Number MFCD00063248
InChI Key TWNIBLMWSKIRAT-UHFFFAOYNA-N
Synonym 1,6-anhydro-beta-d-glucopyranose, levoglucosan, leucoglucosan, 1,6-anhydro-beta-d-glucose, 1,6-anhydro-beta-glucopyranose, glucosan, 1,6-anhydroglucose, unii-5132n17fsd, anhydroglucose, 1,6-anhydro-d-glucose
PubChem CID 2724705
ChEBI CHEBI:30997
IUPAC Name (1R,2S,3S,4R,5R)-6,8-dioxabicyclo[3.2.1]octane-2,3,4-triol
SMILES OC1C2COC(O2)C(O)C1O

Specifications

Melting Point 180°C to 184°C
Quantity 1 g
Beilstein 80998
Solubility Information Soluble in Ethanol and Water.
Formula Weight 162.14
Percent Purity 99%
Chemical Name or Material 1,6-Anhydro-beta-D-glucopyranose

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0448673 Certificate TypeCertificate of Analysis Date04/04/2024 Catalog NumberA16724.01,A16724.03
Lot Number10243334 Certificate TypeCertificate of Analysis Date06/09/2022 Catalog NumberA16724.01,A16724.03
Lot Number10242771 Certificate TypeCertificate of Analysis Date10/08/2022 Catalog NumberA16724.01,A16724.03
Lot Number61500157 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16724.01,A16724.03
Lot Number10170455 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16724.01,A16724.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,6-Anhydro-beta-D-glucopyranose, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.