missing translation for 'onlineSavingsMsg'
Learn More

1-(6-chloro-1,3-benzothiazol-2-yl)hydrazine, 97%, Thermo Scientific™

Product Code. 10526785
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10526785

Brand: Thermo Scientific Maybridge SEW01279DE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 51011-54-2
Molecular Formula C7H6ClN3S
Molecular Weight (g/mol) 199.656
MDL Number MFCD00832729
InChI Key JBAIRAQEQLLZMN-UHFFFAOYSA-N
Synonym 6-chloro-2-hydrazinylbenzo d thiazole, 1-6-chloro-1,3-benzothiazol-2-yl hydrazine, 6-chloro-benzothiazol-2-yl-hydrazine, 6-chloro-2-hydrazinyl-1,3-benzothiazole, 1-6-chlorobenzo d thiazol-2-yl hydrazine, 6-chloro-2-hydrazino-1,3-benzothiazole, 6-chloro-1,3-benzothiazol-2-yl hydrazine, 6-chloro-2-hydrazinobenzothiazole, 2-hydrazino-6-chloro-benzothiazole, 6-chlorobenzothiazole-2-ylhydrazine
PubChem CID 737197
IUPAC Name (6-chloro-1,3-benzothiazol-2-yl)hydrazine
SMILES C1=CC2=C(C=C1Cl)SC(=N2)NN

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 199.66
Percent Purity 97%
Chemical Name or Material 1-(6-Chloro-1, 3-benzothiazol-2-yl)hydrazine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.