missing translation for 'onlineSavingsMsg'
Learn More
1,6-Diaminohexane, 98+%, Thermo Scientific Chemicals Product Code.: 11469080

1,6-Diaminohexane, 98+%, Thermo Scientific Chemicals

Product Code. 11469080
100 g
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11469080

Brand: Thermo Scientific Alfa Aesar A14212.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1,6-Diaminohexane is mainly used as a monomer to make nylon 6-6. Its derivative hexamethylene diisocyanate (HDI) is used in the production of polyurethane. It acts as a cross-linking agent in epoxy resins. Other applications include coatings, lubricants and water treatment products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,6-Diaminohexane is mainly used as a monomer to make nylon 6-6. Its derivative hexamethylene diisocyanate (HDI) is used in the production of polyurethane. It acts as a cross-linking agent in epoxy resins. Other applications include coatings, lubricants and water treatment products.

Solubility
Soluble in water. Slightly soluble in ether, alcohol and benzene.

Notes
Air sensitive and hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids, strong oxidizing agents, acid chlorides, carbon dioxide and acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-09-4
Molecular Formula C6H16N2
MDL Number MFCD00008243
InChI Key NAQMVNRVTILPCV-UHFFFAOYSA-N
Synonym 1,6-hexanediamine, 1,6-diaminohexane, hexamethylenediamine, hmda, 1,6-hexylenediamine, 1,6-hexamethylenediamine, 1,6-diamino-n-hexane, hexamethylene diamine, hexylenediamine, 1,6-hexanediamine solution
PubChem CID 16402
ChEBI CHEBI:39618
IUPAC Name hexane-1,6-diamine

Specifications

Melting Point 39°C to 43°C
Density 0.84
pH 12.4
Boiling Point 199°C to 204°C
Flash Point 85°C (185°F)
Odor Amine-like
Linear Formula H2N(CH2)6NH2
Quantity 100 g
UN Number UN2280
Beilstein 1098307
Sensitivity Air sensitive; Hygroscopic
Merck Index 14,4695
Solubility Information Soluble in water. Slightly soluble in ether,alcohol and benzene.
SMILES C(CCCN)CCN
Molecular Weight (g/mol) 116.208
Formula Weight 116.21
Percent Purity ≥98%
Chemical Name or Material 1,6-Diaminohexane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number62500069 Certificate TypeCertificate of Analysis Date06/02/2025 Catalog NumberA14212.22,A14212.36,A14212-A3,A14212.0E
Lot NumberY13K003 Certificate TypeCertificate of Analysis Date19/11/2024 Catalog NumberA14212.22,A14212.36,A14212-A3
Lot NumberA0461558 Certificate TypeCertificate of Analysis Date18/04/2024 Catalog NumberA14212.22,A14212.36,A14212-A3
Lot NumberA0460105 Certificate TypeCertificate of Analysis Date20/03/2024 Catalog NumberA14212.22,A14212.36,A14212-A3
Lot NumberA0455330 Certificate TypeCertificate of Analysis Date23/02/2024 Catalog NumberA14212.22,A14212.36,A14212-A3
compliance-icons compliance-icons
Product Identifier
  • 1,6-Diaminohexane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 1,6-Hexanediamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,6-Diaminohexane, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.