Learn More
1,6-Dibromohexane, 98%, Thermo Scientific Chemicals
36.58 € - 429.14 €
Chemical Identifiers
CAS | 629-03-8 |
---|---|
Molecular Formula | C6H12Br2 |
Molecular Weight (g/mol) | 243.97 |
MDL Number | MFCD00000272 |
InChI Key | SGRHVVLXEBNBDV-UHFFFAOYSA-N |
Synonym | hexamethylene dibromide, hexane, 1,6-dibromo, 1,6-dibromo-n-hexane, 1,6-dibromohexan, alpha,omega-dibromohexane, 1,6-dibromohexan german, hexamethylene bromide, labotest-bb ltbb001557, 1,6 dibromohexane |
PubChem CID | 12368 |
IUPAC Name | 1,6-dibromohexane |
SMILES | C(CCCBr)CCBr |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10182900
|
Thermo Scientific Acros
112811000 |
100 mL | Glass bottle |
36.58 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10469210
|
Thermo Scientific Acros
112815000 |
500 mL | Glass bottle |
95.07 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11924491
|
Thermo Scientific Acros
112810025 |
2.5 L | Glass bottle |
429.14 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
629-03-8 | |
243.97 | |
SGRHVVLXEBNBDV-UHFFFAOYSA-N | |
12368 | |
C(CCCBr)CCBr |
C6H12Br2 | |
MFCD00000272 | |
hexamethylene dibromide, hexane, 1,6-dibromo, 1,6-dibromo-n-hexane, 1,6-dibromohexan, alpha,omega-dibromohexane, 1,6-dibromohexan german, hexamethylene bromide, labotest-bb ltbb001557, 1,6 dibromohexane | |
1,6-dibromohexane |
Spécification
629-03-8 | |
100.0 | |
Colorless to Yellow | |
243.0°C | |
Authentic | |
C6H12Br2 | |
1.5056 to 1.5076 | |
01,145 | |
hexamethylene dibromide, hexane, 1,6-dibromo, 1,6-dibromo-n-hexane, 1,6-dibromohexan, alpha,omega-dibromohexane, 1,6-dibromohexan german, hexamethylene bromide, labotest-bb ltbb001557, 1,6 dibromohexane | |
SGRHVVLXEBNBDV-UHFFFAOYSA-N | |
1,6-dibromohexane | |
12368 | |
98% | |
1,6-Dibromohexane, 98% |
97.5 | |
-2.0°C | |
1.5800g/mL | |
110°C | |
98% | |
Br(CH2)6Br | |
MFCD00000272 | |
1.58 | |
Solubility in water: insoluble. | |
C(CCCBr)CCBr | |
243.97 | |
243.97 | |
Liquid |
Sécurité et traitement
GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation or rash occurs: Get medic
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 211-067-2
missing translation for 'rtecsNumber' : MO1515000
missing translation for 'tsca' : TSCA
- 1,6-Dibromohexane
Mention d'avertissement
- Warning
Catégorie de danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Respiratory sensitiser Category 1
Mention de danger
- H317-May cause an allergic skin reaction.
- H411-Toxic to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
Conseils de prudence
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P501-Dispose of contents/container to .
RUO – Research Use Only