missing translation for 'onlineSavingsMsg'
Learn More

1,6-Diiodohexane, 97+%, stab. with copper, Thermo Scientific Chemicals

Product Code. 11439150
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1,6-Diiodohexane is used as a precursor in the preparation of cationic tissue plasminogen activator (t-PA)-containing polyurethane surfaces. It is also used as a solvent additive and in potential power conversion efficiency of polymer or fullerene bulk heterojunction solar cells. Further, it is employed in the preparation of 3-(iodohexyl)-1-(3-(triethoxysilyl)propylcarbamoyl)-1H-benzo[d]imidazol-3-ium iodide and 2,4-dihydroxybenzophenone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,6-Diiodohexane is used as a precursor in the preparation of cationic tissue plasminogen activator (t-PA)-containing polyurethane surfaces. It is also used as a solvent additive and in potential power conversion efficiency of polymer or fullerene bulk heterojunction solar cells. Further, it is employed in the preparation of 3-(iodohexyl)-1-(3-(triethoxysilyl)propylcarbamoyl)-1H-benzo[d]imidazol-3-ium iodide and 2,4-dihydroxybenzophenone.

Solubility
Immiscible with water.

Notes
Light sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-09-4
Molecular Formula C6H12I2
Molecular Weight (g/mol) 337.97
MDL Number MFCD00001103
InChI Key QLIMAARBRDAYGQ-UHFFFAOYSA-N
Synonym hexamethylene diiodide, hexane, 1,6-diiodo, 1,6-diiodohexane, stabilized with copper, 1,6-di-iodohexane, 1,6-diiodo hexane, pubchem3846, hexamethyleneiodide, 1,6-bis iodanyl hexane, acmc-209tt7, ksc354g6d
PubChem CID 12373
IUPAC Name 1,6-diiodohexane
SMILES ICCCCCCI

Specifications

Density 2.052
Boiling Point 123°C to 128°C (4 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5852
Quantity 25 g
Beilstein 1280128
Sensitivity Light sensitive
Solubility Information Immiscible with water.
Formula Weight 337.97
Percent Purity 98%
Chemical Name or Material 1,6-Diiodohexane, stabilized with copper
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'