missing translation for 'onlineSavingsMsg'
Learn More

1,6-Diisocyanatohexane, 99+%

Product Code. 10408260 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
25g
This item is not returnable. View return policy

Product Code. 10408260

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 822-06-0 | C8H12N2O2 | 168.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 822-06-0
Molecular Formula C8H12N2O2
Molecular Weight (g/mol) 168.20
MDL Number MFCD00002047
InChI Key RRAMGCGOFNQTLD-UHFFFAOYSA-N
Synonym hexamethylene diisocyanate, 1,6-hexamethylene diisocyanate, hmdi, hexane, 1,6-diisocyanato, hexane 1,6-diisocyanate, 1,6-hexylene diisocyanate, hexamethylene-1,6-diisocyanate, 1,6-hexanediol diisocyanate, hdi, isocyanic acid, hexamethylene ester
PubChem CID 13192
ChEBI CHEBI:53578
IUPAC Name 1,6-diisocyanatohexane
SMILES O=C=NCCCCCCN=C=O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -67°C
Color Colorless to Yellow
Density 1.0400g/mL
Boiling Point 255.0°C
Flash Point 130°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula OCN(CH2)6NCO
Refractive Index 1.4510 to 1.4530
Quantity 100 g
Beilstein 04, II, 711
Specific Gravity 1.04
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in benzene,acetone,toluene,,ethyl acetate,chlorobenzene,very soluble in ether
Formula Weight 168.19
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 1, 6-Diisocyanatohexane, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,6-Diisocyanatohexane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.