missing translation for 'onlineSavingsMsg'
Learn More
1,6-Hexanediamine, 60 wt % aqueous soln., Thermo Scientific Chemicals Product Code.: 10415081

1,6-Hexanediamine, 60 wt % aqueous soln., Thermo Scientific Chemicals

Product Code. 10415081
250 g, Plastic bottle
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10415081

Brand: Thermo Scientific Acros 411562500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-09-4
Molecular Formula C6H16N2
Molecular Weight (g/mol) 116.21
MDL Number MFCD00008243
InChI Key NAQMVNRVTILPCV-UHFFFAOYSA-N
Synonym 1,6-hexanediamine, 1,6-diaminohexane, hexamethylenediamine, hmda, 1,6-hexylenediamine, 1,6-hexamethylenediamine, 1,6-diamino-n-hexane, hexamethylene diamine, hexylenediamine, 1,6-hexanediamine solutionShow MoreShow Less
PubChem CID 16402
ChEBI CHEBI:39618
IUPAC Name hexane-1,6-diamine
SMILES C(CCCN)CCN

Specifications

Density 0.9300g/mL
Flash Point 116°C
Packaging Plastic bottle
Linear Formula NH2(CH2)6NH2
Quantity 250 g
Beilstein 04, 269
Merck Index 15, 4727
Specific Gravity 0.93
Solubility Information Solubility in water: moderately soluble. Other solubilities: soluble in ethanol
Formula Weight 116.21
Physical Form Solution
Chemical Name or Material 1, 6-Hexanediamine, 60 wt % aqueous soln.
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462513 Certificate TypeCertificate of Analysis Date03/05/2024 Catalog Number411560010,411562500
Lot NumberA0456190 Certificate TypeCertificate of Analysis Date25/09/2023 Catalog Number411560010,411562500
Lot NumberA0454875 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number411560010,411562500
Lot NumberA0454875 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number411560010,411562500
Lot NumberA0450999 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number411560010,411562500

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,6-Hexanediamine, 60 wt % aqueous soln., Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.