missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-[6-(Trifluoromethyl)pyridin-2-yl]piperazine, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 231.22 |
Percent Purity | 97% |
Chemical Name or Material | 1-[6-(trifluoromethyl)pyridin-2-yl]piperazine |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
1-[6-(Trifluoromethyl)pyridin-2-yl]piperazine, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction