Learn More
1,8-Diazabicyclo[5.4.0]undec-7-ene, 98+%, Thermo Scientific Chemicals
42.41 € - 252.26 €
Chemical Identifiers
CAS | 6674-22-2 |
---|---|
Molecular Formula | C9H16N2 |
Molecular Weight (g/mol) | 152.241 |
MDL Number | MFCD00006930 |
InChI Key | GQHTUMJGOHRCHB-UHFFFAOYSA-N |
Synonym | 1,8-diazabicyclo 5.4.0 undec-7-ene, 2,3,4,6,7,8,9,10-octahydropyrimido 1,2-a azepine, polycat dbu, dbu, unii-h1ilj6ibux, pyrimido 1,2-a azepine, 2,3,4,6,7,8,9,10-octahydro, 1,8-diazabicyclo 5.4.0-7-undecene, h1ilj6ibux, 1,8-diazabicyclo 5,4,0 undec-7-eneShow More |
PubChem CID | 81184 |
IUPAC Name | 2,3,4,6,7,8,9,10-octahydropyrimido[1,2-a]azepine |
SMILES | C1CCC2=NCCCN2CC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11479673
|
Thermo Scientific Alfa Aesar
A12449.14 |
25 g |
42.41 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11489673
|
Thermo Scientific Alfa Aesar
A12449.22 |
100 g |
106.61 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11499673
|
Thermo Scientific Alfa Aesar
A12449.36 |
500 g |
252.26 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1,8-Diazabicyclo[5.4.0]undec-7-ene is used in organic synthesis as a catalyst, a complexing ligand, and a non-nucleophilic base. It is used as a protecting agent for the synthesis of cephalosporin and as a catalyst for polyurethane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,8-Diazabicyclo[5.4.0]undec-7-ene is used in organic synthesis as a catalyst, a complexing ligand, and a non-nucleophilic base. It is used as a protecting agent for the synthesis of cephalosporin and as a catalyst for polyurethane.
Solubility
Miscible with water.
Notes
Incompatible with acids, strong oxidizing agents, acid anhydrides, acid chlorides and chloroformates.
Chemical Identifiers
6674-22-2 | |
152.241 | |
GQHTUMJGOHRCHB-UHFFFAOYSA-N | |
81184 | |
C1CCC2=NCCCN2CC1 |
C9H16N2 | |
MFCD00006930 | |
1,8-diazabicyclo 5.4.0 undec-7-ene, 2,3,4,6,7,8,9,10-octahydropyrimido 1,2-a azepine, polycat dbu, dbu, unii-h1ilj6ibux, pyrimido 1,2-a azepine, 2,3,4,6,7,8,9,10-octahydro, 1,8-diazabicyclo 5.4.0-7-undecene, h1ilj6ibux, 1,8-diazabicyclo 5,4,0 undec-7-eneShow More | |
2,3,4,6,7,8,9,10-octahydropyrimido[1,2-a]azepine |
Specifications
6674-22-2 | |
1.02 | |
261°C | |
Amine-like | |
1.522 | |
UN3267 | |
Air Sensitive | |
Miscible with water. | |
C1CCC2=NCCCN2CC1 | |
152.241 | |
152.24 | |
1,8-Diazabicyclo[5.4.0]undec-7-ene |
-70°C | |
12.8 | |
116°C (240°F) | |
C9H16N2 | |
MFCD00006930 | |
508906 | |
1,8-diazabicyclo 5.4.0 undec-7-ene, 2,3,4,6,7,8,9,10-octahydropyrimido 1,2-a azepine, polycat dbu, dbu, unii-h1ilj6ibux, pyrimido 1,2-a azepine, 2,3,4,6,7,8,9,10-octahydro, 1,8-diazabicyclo 5.4.0-7-undecene, h1ilj6ibux, 1,8-diazabicyclo 5,4,0 undec-7-ene | |
GQHTUMJGOHRCHB-UHFFFAOYSA-N | |
2,3,4,6,7,8,9,10-octahydropyrimido[1,2-a]azepine | |
81184 | |
99% |
Safety and Handling
GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H301-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 229-713-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
- 1,8-Diazabicyclo[5.4.0]undec-7-ene
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H301-Toxic if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : 2,3,4,6,7,8,9,10-octahydro-pyrimido[1,2-a]azepine
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.