Learn More
1,8-Dibromooctane, 98%, Thermo Scientific Chemicals
48.20 € - 324.00 €
Chemical Identifiers
CAS | 4549-32-0 |
---|---|
Molecular Formula | C8H16Br2 |
MDL Number | MFCD00000277 |
InChI Key | DKEGCUDAFWNSSO-UHFFFAOYSA-N |
Synonym | octamethylene dibromide, octane, 1,8-dibromo, octamethylene bromide, 1,8-dibromo-octane, labotest-bb ltbb002638, 1,8-dibromoctane, octane,8-dibromo, dibromo-1,8 octane, pubchem3905, acmc-209k2r |
PubChem CID | 78310 |
IUPAC Name | 1,8-dibromooctane |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11444210
|
Thermo Scientific Alfa Aesar
A15651.14 |
25 g |
48.20 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11437567
|
Thermo Scientific Alfa Aesar
A15651.18 |
50 g |
94.30 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11454210
|
Thermo Scientific Alfa Aesar
A15651.22 |
100 g |
164.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15453107
|
Thermo Scientific Alfa Aesar
A15651.30 |
250 g |
324.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1,8-Dibromooctane is used as an additive in the preparation poly[[4,8-bis[(2-ethylhexyl)oxy]benzo[1,2-b:4,5-b']dithiophene-2,6-diyl][3-fluoro-2-[(2-ethylhexyl)carbonyl]thieno[3,4-b]thiophenediyl]]:[6,6]-phenylC71-butyric acid methyl ester, which finds application in the fabrication of organic photovoltaics (OPVs). Further, it is used in the preparation of (1,4-bis-(8-bromooctyloxy)-benzene).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,8-Dibromooctane is used as an additive in the preparation poly[[4,8-bis[(2-ethylhexyl)oxy]benzo[1,2-b:4,5-b′]dithiophene-2,6-diyl][3-fluoro-2-[(2-ethylhexyl)carbonyl]thieno[3,4-b]thiophenediyl]]:[6,6]-phenylC71-butyric acid methyl ester, which finds application in the fabrication of organic photovoltaics (OPVs). Further, it is used in the preparation of (1,4-bis-(8-bromooctyloxy)-benzene).
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents and strong bases.
Chemical Identifiers
4549-32-0 | |
MFCD00000277 | |
octamethylene dibromide, octane, 1,8-dibromo, octamethylene bromide, 1,8-dibromo-octane, labotest-bb ltbb002638, 1,8-dibromoctane, octane,8-dibromo, dibromo-1,8 octane, pubchem3905, acmc-209k2r | |
1,8-dibromooctane |
C8H16Br2 | |
DKEGCUDAFWNSSO-UHFFFAOYSA-N | |
78310 |
Specifications
4549-32-0 | |
1.468 | |
>110°C (230°F) | |
1.4995 | |
1698114 | |
Immiscible with water. | |
C(CCCCBr)CCCBr | |
272.024 | |
272.02 | |
1,8-Dibromooctane |
15°C to 16°C | |
270°C to 272°C | |
C8H16Br2 | |
MFCD00000277 | |
octamethylene dibromide, octane, 1,8-dibromo, octamethylene bromide, 1,8-dibromo-octane, labotest-bb ltbb002638, 1,8-dibromoctane, octane,8-dibromo, dibromo-1,8 octane, pubchem3905, acmc-209k2r | |
DKEGCUDAFWNSSO-UHFFFAOYSA-N | |
1,8-dibromooctane | |
78310 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'einecsNumber' : 224-912-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only