missing translation for 'onlineSavingsMsg'
Learn More

1,8-Dichloroanthraquinone, 96%, Thermo Scientific Chemicals

42.11 € - 581.90 €

Chemical Identifiers

CAS 82-43-9
Molecular Formula C14H6Cl2O2
Molecular Weight (g/mol) 277.1
MDL Number MFCD00001191
InChI Key VBQNYYXVDQUKIU-UHFFFAOYSA-N
Synonym 1,8-dichloroanthraquinone, 9,10-anthracenedione, 1,8-dichloro, 1,8-dichloranthrachinon, 1,8-dichloro-9,10-anthraquinone, anthraquinone, 1,8-dichloro, 1,8-dichloranthrachinon czech, acmc-1bmbx, 9, 1,8-dichloro, anthraquinone,8-dichloro
PubChem CID 6708
IUPAC Name 1,8-dichloroanthracene-9,10-dione
SMILES C1=CC2=C(C(=C1)Cl)C(=O)C3=C(C2=O)C=CC=C3Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11495387
View Documents
Thermo Scientific Alfa Aesar
B23421.14
25 g
42.11 €
25g
Please to purchase this item. Need a web account? Register with us today!
11405397
View Documents
Thermo Scientific Alfa Aesar
B23421.22
100 g
131.66 €
100g
Please to purchase this item. Need a web account? Register with us today!
11415397
View Documents
Thermo Scientific Alfa Aesar
B23421.36
500 g
581.90 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

1,8-Dichloroanthraquinone is a reagent in the preparation of anthraquinones, which has antitumor activities. It is used as primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,8-Dichloroanthraquinone is a reagent in the preparation of anthraquinones, which has antitumor activities. It is used as primary and secondary intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agent.
Specifications

Chemical Identifiers

82-43-9
277.1
VBQNYYXVDQUKIU-UHFFFAOYSA-N
6708
C1=CC2=C(C(=C1)Cl)C(=O)C3=C(C2=O)C=CC=C3Cl
C14H6Cl2O2
MFCD00001191
1,8-dichloroanthraquinone, 9,10-anthracenedione, 1,8-dichloro, 1,8-dichloranthrachinon, 1,8-dichloro-9,10-anthraquinone, anthraquinone, 1,8-dichloro, 1,8-dichloranthrachinon czech, acmc-1bmbx, 9, 1,8-dichloro, anthraquinone,8-dichloro
1,8-dichloroanthracene-9,10-dione

Specifications

82-43-9
C14H6Cl2O2
1,8-dichloroanthraquinone, 9,10-anthracenedione, 1,8-dichloro, 1,8-dichloranthrachinon, 1,8-dichloro-9,10-anthraquinone, anthraquinone, 1,8-dichloro, 1,8-dichloranthrachinon czech, acmc-1bmbx, 9, 1,8-dichloro, anthraquinone,8-dichloro
VBQNYYXVDQUKIU-UHFFFAOYSA-N
1,8-dichloroanthracene-9,10-dione
6708
96%
201°C to 203°C
MFCD00001191
Slightly soluble in water.
C1=CC2=C(C(=C1)Cl)C(=O)C3=C(C2=O)C=CC=C3Cl
277.1
277.11
1,8-Dichloroanthraquinone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.

Warning

missing translation for 'einecsNumber' : 201-420-9

missing translation for 'rtecsNumber' : CB6496000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.