Learn More
1,9-Diaminononane, 98%, Thermo Scientific Chemicals
148.00 € - 583.00 €
Chemical Identifiers
CAS | 646-24-2 |
---|---|
Molecular Formula | C9H22N2 |
Molecular Weight (g/mol) | 158.289 |
MDL Number | MFCD00008251 |
InChI Key | SXJVFQLYZSNZBT-UHFFFAOYSA-N |
Synonym | 1,9-diaminononane, 1,9-nonanediamine, nonamethylenediamine, 1,9-diaminonane, nonan-1,9-diamin, 1.9-diaminononane, acmc-1atpv, 1,9-nonamethylenediamine |
PubChem CID | 69534 |
IUPAC Name | nonane-1,9-diamine |
SMILES | C(CCCCN)CCCCN |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11381487
|
Thermo Scientific Alfa Aesar
L02043.06 |
5 g |
148.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11391487
|
Thermo Scientific Alfa Aesar
L02043.14 |
25 g |
583.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1,9-Diaminononane is used as an organic intermediate. It is also used to detect spermine and spermidine by matrix-assisted laser desorption/ionization combined with time-of-flight mass spectrometry ( MALDI-TOF MS) in foods and for the identification of apoptosis-related proteins by nano-flow ultra-performance liquid chromatography tandem mass-spectrometry (UPLC-MS/MS) after treatment with spermine and spermidine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,9-Diaminononane is used as an organic intermediate. It is also used to detect spermine and spermidine by matrix-assisted laser desorption/ionization combined with time-of-flight mass spectrometry ( MALDI-TOF MS) in foods and for the identification of apoptosis-related proteins by nano-flow ultra-performance liquid chromatography tandem mass-spectrometry (UPLC-MS/MS) after treatment with spermine and spermidine.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with acids, acid chlorides, acid anhydrides, oxidizing agents and carbon dioxide.
Chemical Identifiers
646-24-2 | |
158.289 | |
SXJVFQLYZSNZBT-UHFFFAOYSA-N | |
69534 | |
C(CCCCN)CCCCN |
C9H22N2 | |
MFCD00008251 | |
1,9-diaminononane, 1,9-nonanediamine, nonamethylenediamine, 1,9-diaminonane, nonan-1,9-diamin, 1.9-diaminononane, acmc-1atpv, 1,9-nonamethylenediamine | |
nonane-1,9-diamine |
Specifications
646-24-2 | |
258°C to 259°C | |
C9H22N2 | |
UN3259 | |
Air Sensitive | |
Soluble in water. | |
C(CCCCN)CCCCN | |
158.289 | |
158.29 | |
1,9-Diaminononane |
36°C to 40°C | |
>110°C (230°F) | |
MFCD00008251 | |
1737533 | |
1,9-diaminononane, 1,9-nonanediamine, nonamethylenediamine, 1,9-diaminonane, nonan-1,9-diamin, 1.9-diaminononane, acmc-1atpv, 1,9-nonamethylenediamine | |
SXJVFQLYZSNZBT-UHFFFAOYSA-N | |
nonane-1,9-diamine | |
69534 | |
98% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: POLYAMINES, SOLID, CORROSIVE, N.O.S.
missing translation for 'einecsNumber' : 211-470-3
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only