missing translation for 'onlineSavingsMsg'
Learn More

1-Acetylnaphthalene, 97+%, Thermo Scientific Chemicals

Product Code. 11499733
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

They are used as ingredients of fragrances. Their derivatives are used as anti-tubercular agents. The end applications include soap, detergent, beauty care product, household product. It is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
They are used as ingredients of fragrances. Their derivatives are used as anti-tubercular agents. The end applications include soap, detergent, beauty care product, household product. It is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products.

Solubility
Not miscible or difficult to mix in water. Soluble in ethyl alcohol. Insoluble in paraffin oil.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 941-98-0
Molecular Formula C12H10O
Molecular Weight (g/mol) 170.211
MDL Number MFCD00004013
InChI Key QQLIGMASAVJVON-UHFFFAOYSA-N
Synonym 1'-acetonaphthone, 1-acetonaphthone, 1-acetylnaphthalene, methyl 1-naphthyl ketone, 1-acetonaphthalene, 1-1-naphthalenyl ethanone, ethanone, 1-1-naphthalenyl, 1-naphthalen-1-yl ethanone, 1-naphthyl methyl ketone, alpha-acetonaphthone
PubChem CID 13663
IUPAC Name 1-naphthalen-1-ylethanone
SMILES CC(=O)C1=CC=CC2=CC=CC=C21

Specifications

Melting Point 9°C to 11°C
Density 1.12
Boiling Point 302°C
Flash Point >110°C (230°F)
Refractive Index 1.628
Quantity 250 g
Beilstein 1100618
Solubility Information Not miscible or difficult to mix in water. Soluble in ethyl alcohol. Insoluble in paraffin oil.
Formula Weight 170.21
Percent Purity ≥97%
Chemical Name or Material 1-Acetylnaphthalene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'