missing translation for 'onlineSavingsMsg'
Learn More

1-Acetylpyrene, 97%

Product Code. 15453247
2.5 g
Click to view available options
Quantity:
2.5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 3264-21-9 | C18H12O | 244.293 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 3264-21-9
Melting Point 86°C to 89°C
Molecular Formula C18H12O
MDL Number MFCD00021663
Quantity 2.5 g
Synonym 1-acetylpyrene, ethanone, 1-1-pyrenyl, 1-pyren-1-yl ethanone, ethanone,1-1-pyrenyl, acetylpyrene, 1-1-pyrenyl ethanone #, acmc-1cs50
InChI Key KCIJNJVCFPSUBQ-UHFFFAOYSA-N
SMILES CC(=O)C1=C2C=CC3=CC=CC4=C3C2=C(C=C4)C=C1
IUPAC Name 1-pyren-1-ylethanone
Molecular Weight (g/mol) 244.293
PubChem CID 96251
Formula Weight 244.29
Percent Purity 97%
Chemical Name or Material 1-Acetylpyrene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Acetylpyrene, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.