Learn More
1-Adamantanamine hydrochloride, 99%, Thermo Scientific Chemicals
50.35 € - 657.27 €
Chemical Identifiers
CAS | 665-66-7 |
---|---|
Molecular Formula | C10H18ClN |
Molecular Weight (g/mol) | 187.71 |
MDL Number | MFCD00074723 |
InChI Key | WOLHOYHSEKDWQH-UHFFFAOYSA-N |
Synonym | amantadine hydrochloride, 1-adamantanamine hydrochloride, symmetrel, 1-aminoadamantane hydrochloride, amantadine hcl, adamantanamine hydrochloride, amazolon, midantan, midantane, mydantane |
PubChem CID | 64150 |
ChEBI | CHEBI:2619 |
IUPAC Name | adamantan-1-amine;hydrochloride |
SMILES | [H+].[Cl-].NC12CC3CC(CC(C3)C1)C2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11410613
|
Thermo Scientific Alfa Aesar
A12699.14 |
25 g |
50.35 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11420613
|
Thermo Scientific Alfa Aesar
A12699.22 |
100 g |
171.86 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11430613
|
Thermo Scientific Alfa Aesar
A12699.36 |
500 g |
657.27 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-Adamantanamine hydrochloride is used in the prophylactic or symptomatic treatment of influenza A. It is also used as an antiparkinsonian agent, to treat extra pyramidal reactions, and for postherpetic neuralgia. It is also used an NMDA-receptor antagoinst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Adamantanamine hydrochloride is used in the prophylactic or symptomatic treatment of influenza A. It is also used as an antiparkinsonian agent, to treat extra pyramidal reactions, and for postherpetic neuralgia. It is also used an NMDA-receptor antagoinst.
Solubility
Soluble in water (50 mg/ml), ethanol (38 mg/ml at 25°C), chloroform (1:18), DMSO (38 mg/ml at 25°C), and methanol
Notes
Hygroscopic. Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent and water/moisture.
Chemical Identifiers
665-66-7 | |
187.71 | |
WOLHOYHSEKDWQH-UHFFFAOYSA-N | |
64150 | |
adamantan-1-amine;hydrochloride |
C10H18ClN | |
MFCD00074723 | |
amantadine hydrochloride, 1-adamantanamine hydrochloride, symmetrel, 1-aminoadamantane hydrochloride, amantadine hcl, adamantanamine hydrochloride, amazolon, midantan, midantane, mydantane | |
CHEBI:2619 | |
[H+].[Cl-].NC12CC3CC(CC(C3)C1)C2 |
Specifications
665-66-7 | |
C10H18ClN | |
4198854 | |
amantadine hydrochloride, 1-adamantanamine hydrochloride, symmetrel, 1-aminoadamantane hydrochloride, amantadine hcl, adamantanamine hydrochloride, amazolon, midantan, midantane, mydantane | |
WOLHOYHSEKDWQH-UHFFFAOYSA-N | |
adamantan-1-amine;hydrochloride | |
64150 | |
187.71 | |
1-Adamantanamine hydrochloride |
360°C | |
MFCD00074723 | |
14374 | |
Soluble in water (50mg/ml),ethanol (38mg/ml at 25°C),chloroform (1:18),DMSO (38mg/ml at 25°C),and methanol | |
[H+].[Cl-].NC12CC3CC(CC(C3)C1)C2 | |
187.71 | |
CHEBI:2619 | |
99% |
Safety and Handling
Harmful if swallowed
Acute toxicity (category 4)
missing translation for 'einecsNumber' : 211-560-2
missing translation for 'rtecsNumber' : AU4375000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.