missing translation for 'onlineSavingsMsg'
Learn More

1-Adamantanamine hydrochloride, 99+%, Thermo Scientific Chemicals

Codice prodotto. 10645972
Click to view available options
Quantity:
25 g
100 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Adamantanamine hydrochloride is used in the prophylactic or symptomatic treatment of influenza A. It is also used as an antiparkinsonian agent, to treat extra pyramidal reactions, and for postherpetic neuralgia. It is also used an NMDA-receptor antagoinst.

Solubility
Soluble in water (50 mg/ml), ethanol (38 mg/ml at 25°C), chloroform (1:18), DMSO (38 mg/ml at 25°C), and methanol

Notes
Hygroscopic. Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent and water/moisture.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 665-66-7
Molecular Formula C10H18ClN
Molecular Weight (g/mol) 187.71
MDL Number MFCD00074723
InChI Key WOLHOYHSEKDWQH-UHFFFAOYSA-N
Synonym amantadine hydrochloride, 1-adamantanamine hydrochloride, symmetrel, 1-aminoadamantane hydrochloride, amantadine hcl, adamantanamine hydrochloride, amazolon, midantan, midantane, mydantane
PubChem CID 64150
ChEBI CHEBI:2619
IUPAC Name adamantan-1-amine;hydrochloride
SMILES [H+].[Cl-].NC12CC3CC(CC(C3)C1)C2

Specifica

CAS Min % 99
CAS Max % 100
Melting Point >300.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 25 g
Merck Index 15, 368
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and chloroform,practically insoluble in ether
Formula Weight 187.71
Percent Purity 99+%
Physical Form Fine Crystalline Powder
Chemical Name or Material 1-Adamantanamine hydrochloride, 99+%
Vedi altri risultati Mostra meno risultati
compliance-icons
Identificativo prodotto
  • 1-Adamantanamine hydrochloride
Avvertenza
  • Warning
Categoria di pericolo
  • Acute toxicity Category 4
Indicazioni di pericolo
  • H302-Harmful if swallowed.
Consigli di prudenza
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'