missing translation for 'onlineSavingsMsg'
Learn More

1-Amino-1-cyclopropanecarbonitrile hydrochloride, 97%, Thermo Scientific Chemicals

Product Code. 11322349
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127946-77-4
Molecular Formula C4H7ClN2
Molecular Weight (g/mol) 118.564
MDL Number MFCD04114063
InChI Key PCEIEQLJYDMRFZ-UHFFFAOYSA-N
Synonym 1-amino-1-cyclopropanecarbonitrile hydrochloride, 1-aminocyclopropanecarbonitrile hydrochloride, 1-aminocyclopropane-1-carbonitrile hydrochloride, 1-aminocyclopropanecarbonitrile hcl, 1-amino-cyclopropyl cyanic hydrochloride, 1-amino-1-cyanocyclopropane hydrochloride, 1-amino-cyclopropanecarbonitrile hydrochloride, cyclopropanecarbonitrile, 1-amino-, monohydrochloride, 1-cyanocyclopropanaminium chloride, pubchem16832
PubChem CID 14586363
IUPAC Name 1-aminocyclopropane-1-carbonitrile;hydrochloride
SMILES C1CC1(C#N)N.Cl

Specifications

Melting Point ∽220°C (decomposition)
Quantity 5 g
UN Number UN3439
Formula Weight 118.56
Percent Purity 97%
Chemical Name or Material 1-Amino-1-cyclopropanecarbonitrile hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.