missing translation for 'onlineSavingsMsg'
Learn More

1-Amino-3-(4-methoxyphenoxy)propan-2-ol, ≥95%, Thermo Scientific™

Produktcode. 10763033
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemische identificatiekenmerken

CAS 5002-93-7
Molecular Formula C10H15NO3
Molecular Weight (g/mol) 197.234
MDL Number MFCD00205693
InChI Key KFQPRNVTVMCYEH-UHFFFAOYSA-N
Synonym 1-amino-3-4-methoxyphenoxy propan-2-ol, 1-amino-3-4-methoxy-phenoxy-propan-2-ol, 2-propanol, 1-amino-3-4-methoxyphenoxy, 3-amino-1-4-methoxyphenoxy propan-2-ol, maybridge1_002199, 4-3-amino-2-hydroxypropoxy anisole, 2rs-1-amino-3-4methoxyphenoxy-2-propanol, 2rs-1-amino-3-4-methoxyphenoxy-2-propanol
PubChem CID 2735360
IUPAC Name 1-amino-3-(4-methoxyphenoxy)propan-2-ol
SMILES COC1=CC=C(C=C1)OCC(CN)O

Specificatie

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95+%
Packaging Amber Glass Bottle
Linear Formula CH3OC6H4OCH2CH(OH)CH2NH2
Quantity 1 g
Formula Weight 197.23
Percent Purity 95%
Chemical Name or Material 1-Amino-3-(4-methoxyphenoxy)propan-2-ol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.