missing translation for 'onlineSavingsMsg'
Learn More

1-Aza-18-crown-6, 95%, Thermo Scientific Chemicals

Product Code. 11309286
Click to view available options
Quantity:
250 mg
1 g
5 g

1-Aza-18-crown-6 is used as building block for the synthesis of bis-crown ethers, lariat ethers and other monofunctionalized macrocycles. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Aza-18-crown-6 is used as building block for the synthesis of bis-crown ethers, lariat ethers and other monofunctionalized macrocycles. It is also used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Air sensitive. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33941-15-0
Molecular Formula C12H25NO5
Molecular Weight (g/mol) 263.334
MDL Number MFCD00075466
InChI Key NBXKUSNBCPPKRA-UHFFFAOYSA-N
PubChem CID 118578
IUPAC Name 1,4,7,10,13-pentaoxa-16-azacyclooctadecane
SMILES C1COCCOCCOCCOCCOCCN1

Specifications

Melting Point 46°C to 49°C
Quantity 1 g
Beilstein 1074079
Sensitivity Air Sensitive
Synonym 1-aza-18-crown-6, 1-aza-18-crown6-ether, aza-18-crown-6, acmc-209tw0, 1-aza-18-crown 6-ether, 1-aza-18-crown nt, 1-aza-4,7,10,13,16-pentaoxacyclooctadecane
Solubility Information Insoluble in water.
Formula Weight 263.33
Percent Purity 95%
Chemical Name or Material 1-Aza-18-crown-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.