missing translation for 'onlineSavingsMsg'
Learn More

1-Benzhydryl-3-azetidinyl methanesulfonate, 95%, Thermo Scientific™

Product Code. 15433736
Click to view available options
Quantity:
250 mg
500 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33301-41-6
Molecular Formula C17H19NO3S
Molecular Weight (g/mol) 317.40
MDL Number MFCD00159216
InChI Key MSVZMUILYMLJCF-UHFFFAOYSA-N
Synonym 1-benzhydryl-3-methanesulfonyloxy azetidine, 1-benzhydryl-3-methanesulfonatoazetidine, 1-diphenylmethyl-3-methanesulfonyloxy azetidine, 1-diphenylmethyl-3-azetidinyl methanesulfonate, 1-diphenylmethyl azetidin-3-yl methanesulfonate, 1-benzhydryl-3-azetidinyl methanesulfonate, 1-benzhydrylazetidin-3-yl methanesulfonate, methanesulfonic acid 1-benzhydryl-azetidin-3-yl ester, 1-benzhydryl-3-methanesulfonyloxyazetidine
PubChem CID 2758716
IUPAC Name (1-benzhydrylazetidin-3-yl) methanesulfonate
SMILES CS(=O)(=O)OC1CN(C1)C(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 111°C to 112°C
Quantity 500 mg
Formula Weight 317.41
Percent Purity 95%
Chemical Name or Material 1-Benzhydryl-3-azetidinyl methanesulfonate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'