missing translation for 'onlineSavingsMsg'
Learn More

1-Benzofuran-5-ylmethanol, Thermo Scientific™

Product Code. 10285164
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 31823-05-9
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.16
MDL Number MFCD04115373
InChI Key XSLXZYDPOMAXTM-UHFFFAOYSA-N
Synonym benzofuran-5-ylmethanol, 5-benzofuranmethanol, 5-hydroxymethylbenzofuran, pubchem7026, 5-hydroxymethyl-benzofuran, 5-hydroxymethylbenzo b furan, benzofuran-5-methanol, 1-benzofuran-5-yl methanol, 1-benzo b furan-5-ylmethanol, benzo b furan-5-ylmethan-1-ol
PubChem CID 2795179
IUPAC Name 1-benzofuran-5-ylmethanol
SMILES OCC1=CC=C2OC=CC2=C1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 148.16
Percent Purity 97%
Chemical Name or Material 1-Benzofuran-5-ylmethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.