missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Benzothiophen-3-ylmethanol, 97%, Thermo Scientific™
93.40 € - 361.00 €
Chemical Identifiers
CAS | 5381-24-8 |
---|---|
Molecular Formula | C9H8OS |
Molecular Weight (g/mol) | 164.22 |
MDL Number | MFCD02682007 |
InChI Key | UYGMKSKKGSUAHB-UHFFFAOYSA-N |
Synonym | benzo b thiophen-3-ylmethanol, benzo b thiophene-3-methanol, benzothien-3-ylmethanol, benzo, a thiophene-3-methanol, benzo b thiophen-3-yl methanol, 1-benzothiophen-3-yl methanol, 3-hydroxymethylbenzo b thiophene, benzo b thiophen-3-ylmethan-1-ol, 3-hydroxymethyl benzo b thiophene |
PubChem CID | 2776342 |
SMILES | OCC1=CSC2=CC=CC=C12 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10581682
|
Thermo Scientific Maybridge
CC12309DA |
1 g |
93.40 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10215224
|
Thermo Scientific Maybridge
CC12309EA |
10 g |
361.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
5381-24-8 | |
164.22 | |
UYGMKSKKGSUAHB-UHFFFAOYSA-N | |
2776342 |
C9H8OS | |
MFCD02682007 | |
benzo b thiophen-3-ylmethanol, benzo b thiophene-3-methanol, benzothien-3-ylmethanol, benzo, a thiophene-3-methanol, benzo b thiophen-3-yl methanol, 1-benzothiophen-3-yl methanol, 3-hydroxymethylbenzo b thiophene, benzo b thiophen-3-ylmethan-1-ol, 3-hydroxymethyl benzo b thiophene | |
OCC1=CSC2=CC=CC=C12 |
Specifications
5381-24-8 | |
100.0 | |
Amber glass bottle | |
MFCD02682007 | |
UYGMKSKKGSUAHB-UHFFFAOYSA-N | |
1-benzothiophen-3-ylmethanol | |
2776342 | |
97% |
96.5 | |
97% | |
C9H8OS | |
benzo b thiophen-3-ylmethanol, benzo b thiophene-3-methanol, benzothien-3-ylmethanol, benzo, a thiophene-3-methanol, benzo b thiophen-3-yl methanol, 1-benzothiophen-3-yl methanol, 3-hydroxymethylbenzo b thiophene, benzo b thiophen-3-ylmethan-1-ol, 3-hydroxymethyl benzo b thiophene | |
OCC1=CSC2=CC=CC=C12 | |
164.22 | |
164.23 | |
1-Benzothiophen-3-ylmethanol |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title