missing translation for 'onlineSavingsMsg'
Learn More

1-Benzothiophen-5-yl isocyanate, 97%, Thermo Scientific™

Product Code. 10771114
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 239097-78-0
Molecular Formula C9H5NOS
Molecular Weight (g/mol) 175.205
InChI Key GWLZJAWTWVSHCU-UHFFFAOYSA-N
Synonym 1-benzothiophen-5-yl isocyanate, 1-benzothiophen-5-ylisocyanate, benzo b thiophen-5-isocyanate, 5-isocyanato-benzo b thiophene, benzo b thiophene,5-isocyanato
PubChem CID 7060535
IUPAC Name 5-isocyanato-1-benzothiophene
SMILES C1=CC2=C(C=CS2)C=C1N=C=O

Specifications

Assay Percent Range 97%
Quantity 250 mg
Formula Weight 175.21
Percent Purity 97%
Chemical Name or Material 1-Benzothiophen-5-yl isocyanate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'