missing translation for 'onlineSavingsMsg'
Learn More

1-Benzothiophene-3-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10051664
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 5381-25-9
Molecular Formula C9H6O2S
Molecular Weight (g/mol) 178.205
MDL Number MFCD01846406
InChI Key DRBLTQNCQJXSNU-UHFFFAOYSA-N
Synonym benzo b thiophene-3-carboxylic acid, benzothiophene-3-carboxylic acid, benzothiophene-3-carboxylicacid, pubchem13463, acmc-20a10z, ksc178m8d, 1-benzo b thiophene-3-carboxylic acid, #, thianaphthene-3-carboxylic acid
PubChem CID 601280
IUPAC Name 1-benzothiophene-3-carboxylic acid
SMILES C1=CC=C2C(=C1)C(=CS2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 178.21
Percent Purity 97%
Chemical Name or Material 1-Benzothiophene-3-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'