missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-4-hydroxypiperidine, 97%, Thermo Scientific™

Product Code. 11474333
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4727-72-4
Molecular Formula C12H17NO
Molecular Weight (g/mol) 191.274
MDL Number MFCD00006503
InChI Key BPPZXJZYCOETDA-UHFFFAOYSA-N
Synonym 1-benzyl-4-hydroxypiperidine, 1-benzyl-4-piperidinol, n-benzyl-4-hydroxypiperidine, 1-benzyl-piperidin-4-ol, 4-piperidinol, 1-phenylmethyl, n-benzyl-4-hydroxy piperidine, n-benzyl-4-piperidinol, 1-benzyl-4-hydroxy piperidine, 1-phenylmethyl piperidin-4-ol, 1-phenylmethyl-4-piperidinol
PubChem CID 78461
IUPAC Name 1-benzylpiperidin-4-ol
SMILES C1CN(CCC1O)CC2=CC=CC=C2

Specifications

Melting Point 60°C to 64°C
Boiling Point 121°C to 123°C (0.7 mmHg)
Quantity 5 g
Beilstein 146118
Sensitivity Hygroscopic
Formula Weight 191.27
Percent Purity 97%
Chemical Name or Material 1-Benzyl-4-hydroxypiperidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'