Learn More
1-(Benzyloxycarbonyl)piperazine, 98%, Thermo Scientific Chemicals
63.82 € - 689.43 €
Chemical Identifiers
CAS | 31166-44-6 |
---|---|
Molecular Formula | C12H17N2O2 |
Molecular Weight (g/mol) | 221.28 |
MDL Number | MFCD00274317 |
InChI Key | CTOUWUYDDUSBQE-UHFFFAOYSA-O |
Synonym | 1-cbz-piperazine, benzyl 1-piperazinecarboxylate, 1-benzyloxycarbonyl piperazine, 1-carbobenzoxypiperazine, n-cbz-piperazine, 1-z-piperazine, 1-piperazinecarboxylic acid, phenylmethyl ester, piperazine-1-carboxylic acid benzyl ester, phenylmethyl piperazinecarboxylate, cbz-piperazine |
PubChem CID | 643495 |
IUPAC Name | benzyl piperazine-1-carboxylate |
SMILES | O=C(OCC1=CC=CC=C1)N1CC[NH2+]CC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15432428
|
Thermo Scientific Alfa Aesar
H56008.AA |
5 mL |
63.82 €
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15442428
|
Thermo Scientific Alfa Aesar
H56008.AC |
25 mL |
252.26 €
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15452428
|
Thermo Scientific Alfa Aesar
H56008.AE |
100 mL |
689.43 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
31166-44-6 | |
221.28 | |
CTOUWUYDDUSBQE-UHFFFAOYSA-O | |
643495 | |
O=C(OCC1=CC=CC=C1)N1CC[NH2+]CC1 |
C12H17N2O2 | |
MFCD00274317 | |
1-cbz-piperazine, benzyl 1-piperazinecarboxylate, 1-benzyloxycarbonyl piperazine, 1-carbobenzoxypiperazine, n-cbz-piperazine, 1-z-piperazine, 1-piperazinecarboxylic acid, phenylmethyl ester, piperazine-1-carboxylic acid benzyl ester, phenylmethyl piperazinecarboxylate, cbz-piperazine | |
benzyl piperazine-1-carboxylate |
Specifications
31166-44-6 | |
158°C to 161°C (1.4 mmHg) | |
1.546 | |
179500 | |
CTOUWUYDDUSBQE-UHFFFAOYSA-O | |
benzyl piperazine-1-carboxylate | |
643495 | |
98% |
1.142 | |
C12H17N2O2 | |
MFCD00274317 | |
1-cbz-piperazine, benzyl 1-piperazinecarboxylate, 1-benzyloxycarbonyl piperazine, 1-carbobenzoxypiperazine, n-cbz-piperazine, 1-z-piperazine, 1-piperazinecarboxylic acid, phenylmethyl ester, piperazine-1-carboxylic acid benzyl ester, phenylmethyl piperazinecarboxylate, cbz-piperazine | |
O=C(OCC1=CC=CC=C1)N1CC[NH2+]CC1 | |
221.28 | |
220.27 | |
1-(Benzyloxycarbonyl)piperazine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.