missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-BOC-3-(bromomethyl)azetidine, 95%, Thermo Scientific Chemicals
205.00 € - 748.00 €
Chemical Identifiers
CAS | 253176-93-1 |
---|---|
Molecular Formula | C9H16BrNO2 |
Molecular Weight (g/mol) | 250.14 |
MDL Number | MFCD16556174 |
InChI Key | PUKCUGDJEPVLPR-UHFFFAOYSA-N |
Synonym | 1-boc-3-bromomethyl azetidine, tert-butyl 3-bromomethyl azetidine-1-carboxylate, 1-boc-3-bromomethylazetidine, n-tert-butoxycarbonyl-3-bromomethylazetidine, 3-bromomethyl-azetidine-1-carboxylic acid tert-butyl ester, acmc-209d8t, 1-n-boc-3-bromomethyl azetidine, tert-butyl 3-bromomethyl-1-azetidinecarboxylate, tert-butyl 3-bromomethyl ;azetidine-1-carboxylate, 2-methyl-2-propanyl 3-bromomethyl-1-azetidinecarboxylate |
PubChem CID | 53350331 |
IUPAC Name | tert-butyl 3-(bromomethyl)azetidine-1-carboxylate |
SMILES | CC(C)(C)OC(=O)N1CC(CBr)C1 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
253176-93-1 | |
250.14 | |
PUKCUGDJEPVLPR-UHFFFAOYSA-N | |
53350331 | |
CC(C)(C)OC(=O)N1CC(CBr)C1 |
C9H16BrNO2 | |
MFCD16556174 | |
1-boc-3-bromomethyl azetidine, tert-butyl 3-bromomethyl azetidine-1-carboxylate, 1-boc-3-bromomethylazetidine, n-tert-butoxycarbonyl-3-bromomethylazetidine, 3-bromomethyl-azetidine-1-carboxylic acid tert-butyl ester, acmc-209d8t, 1-n-boc-3-bromomethyl azetidine, tert-butyl 3-bromomethyl-1-azetidinecarboxylate, tert-butyl 3-bromomethyl ;azetidine-1-carboxylate, 2-methyl-2-propanyl 3-bromomethyl-1-azetidinecarboxylate | |
tert-butyl 3-(bromomethyl)azetidine-1-carboxylate |
Specifications
253176-93-1 | |
Glass Bottle | |
MFCD16556174 | |
PUKCUGDJEPVLPR-UHFFFAOYSA-N | |
tert-butyl 3-(bromomethyl)azetidine-1-carboxylate | |
53350331 | |
95% |
94% min. (GC) | |
C9H16BrNO2 | |
1-boc-3-bromomethyl azetidine, tert-butyl 3-bromomethyl azetidine-1-carboxylate, 1-boc-3-bromomethylazetidine, n-tert-butoxycarbonyl-3-bromomethylazetidine, 3-bromomethyl-azetidine-1-carboxylic acid tert-butyl ester, acmc-209d8t, 1-n-boc-3-bromomethyl azetidine, tert-butyl 3-bromomethyl-1-azetidinecarboxylate, tert-butyl 3-bromomethyl ;azetidine-1-carboxylate, 2-methyl-2-propanyl 3-bromomethyl-1-azetidinecarboxylate | |
CC(C)(C)OC(=O)N1CC(CBr)C1 | |
250.14 | |
250.13 | |
1-BOC-3-(bromomethyl)azetidine |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 1-Boc-3-(bromomethyl)azetidine, 95%
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only