missing translation for 'onlineSavingsMsg'
Learn More

1-BOC-3-iodoazetidine, 95%, Thermo Scientific Chemicals

45.41 € - 173.00 €

Chemical Identifiers

CAS 254454-54-1
Molecular Formula C8H14INO2
Molecular Weight (g/mol) 283.11
MDL Number MFCD09752821
InChI Key XPDIKRMPZNLBAC-UHFFFAOYSA-N
Synonym 1-boc-3-iodoazetidine, 1-boc-3-iodo-azetidine, 1-boc-3-iodo azetidine, n-boc-3-iodoazetidine, 3-iodo-azetidine-1-carboxylic acid tert-butyl ester, 3-iodoazetidine, n-boc protected, 1-n-boc-3-iodoazetidine, 1-tert-butoxycarbonyl-3-iodoazetidine, tert-butyl 3-iodo-1-azetidinecarboxylate, tert-butyl 3-iodoazetidinecarboxylate
PubChem CID 11000522
IUPAC Name tert-butyl 3-iodoazetidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(I)C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15289626
View Documents
Thermo Scientific Acros
452080010
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15279726
View Documents
Thermo Scientific Acros
452080050
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

254454-54-1
283.11
XPDIKRMPZNLBAC-UHFFFAOYSA-N
11000522
CC(C)(C)OC(=O)N1CC(I)C1
C8H14INO2
MFCD09752821
1-boc-3-iodoazetidine, 1-boc-3-iodo-azetidine, 1-boc-3-iodo azetidine, n-boc-3-iodoazetidine, 3-iodo-azetidine-1-carboxylic acid tert-butyl ester, 3-iodoazetidine, n-boc protected, 1-n-boc-3-iodoazetidine, 1-tert-butoxycarbonyl-3-iodoazetidine, tert-butyl 3-iodo-1-azetidinecarboxylate, tert-butyl 3-iodoazetidinecarboxylate
tert-butyl 3-iodoazetidine-1-carboxylate

Specifications

254454-54-1
Glass Bottle
MFCD09752821
XPDIKRMPZNLBAC-UHFFFAOYSA-N
tert-butyl 3-iodoazetidine-1-carboxylate
11000522
95%
94% min. (GC)
C8H14INO2
1-boc-3-iodoazetidine, 1-boc-3-iodo-azetidine, 1-boc-3-iodo azetidine, n-boc-3-iodoazetidine, 3-iodo-azetidine-1-carboxylic acid tert-butyl ester, 3-iodoazetidine, n-boc protected, 1-n-boc-3-iodoazetidine, 1-tert-butoxycarbonyl-3-iodoazetidine, tert-butyl 3-iodo-1-azetidinecarboxylate, tert-butyl 3-iodoazetidinecarboxylate
CC(C)(C)OC(=O)N1CC(I)C1
283.11
283.1
1-BOC-3-iodoazetidine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS Signal Word: Warning

Product Identifier
  • 1-Boc-3-iodoazetidine, 95%

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.